Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trophinin Mouse anti-Human, Mouse, Clone: 3-11, MilliporeSigma™

Catalog No. 05946MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-946-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-946-MI Supplier MilliporeSigma Supplier No. 05946
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Trophinin Antibody, clone 3-11 detects level of Trophinin and has been published and validated for use in immunoprecipitation, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Trophinin
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3-11
Conjugate Unconjugated
Gene Accession No. NM_177555
Gene Symbols TRO, KIAA1114, MAGE-d3, MAGE-D3, Trophinin, MAGED3, magphinin, trophinin
Host Species Mouse
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 1319-1327 (SIVGFSGGP-C) of human Trophinin (MAGE-D3)
Purification Method Cell culture supernatant
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C for 24 months.
Form Supernatant
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.