Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tropomyosin-5 Mouse anti-Mouse, Human, Rat, Clone: 2G10.2, MilliporeSigma™

Catalog No. MAB2256MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2256MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2256MI Supplier MilliporeSigma Supplier No. MAB2256

Mouse Monoclonal Antibody

Anti-Tropomyosin-5 Antibody, clone 2G10.2 is an antibody against Tropomyosin-5 for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tropomyosin-5
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2G10.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_476556
Gene Symbols TPM5P, Tpm5
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to rat Tropomysosin-5.
Purification Method Protein G
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.