Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tropomyosin Sheep anti-Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB5447MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5447MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5447MI Supplier MilliporeSigma Supplier No. AB5447
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Detect Tropomyosin using this Anti-Tropomyosin Antibody, 5NM1 and 5NM2 validated for use in western blot, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tropomyosin
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin in PBS (liquid).
Gene Symbols TPM3; TM30; MGC3261; TRK; NEM1; TM30nm; FLJ41118; Tropomyosin-3; OK/SW-cl.5; TM-5; hscp30; TPMsk3; MGC14582; hTM5; TM3; MGC72094
Host Species Sheep
Immunogen Synthetic peptide corresponding to the 9d exon from the gamma gene of tropomyosin.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_001043351.1; NM_001043352.1; NM_001043353.1; NM_152263.2; NM_153649.3
Target Species Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.