Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tropomyosin Sheep anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB5437SMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5437SMI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5437SMI Supplier MilliporeSigma Supplier No. AB5437S
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Anti-Tropomyosin Antibody, Smooth Muscle detects level of Tropomyosin and has been published and validated for use in western blot, immunocytochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tropomyosin
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid
Gene Symbols TPM1; CMH3; Tropomyosin-1; HTM-alpha; Alpha-tropomyosin; TMSA; C15orf13
Host Species Sheep
Immunogen Synthetic peptide corresponding to the 2a exon from the α gene of tropomyosin.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000366.5; NM_001018004.1; NM_001018005.1; NM_001018006.1; NM_001018007.1; NM_001018008.1; NM_001018020.1
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.