Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tropomyosin Mouse anti-Mouse, Human, Rat, Clone: 15D12.2, MilliporeSigma™

Catalog No. MAB2254MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2254MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2254MI Supplier MilliporeSigma Supplier No. MAB2254
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Tropomyosin using this Anti-Tropomyosin Antibody, clone 15D12.2 validated for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tropomyosin
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 15D12.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine, pH 7.4, 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001029243
Gene Symbols TPM1, TMSA, C15orf13, CMD1Y, CMH3
Host Species Mouse
Immunogen Linear peptide corresponding to the entire exon 9d of rat Tropomyosin.
Purification Method Protein G
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.