Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Troponin I, a.a. 87-93, Clone: C5, ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB1691/DEL

Description
ELISA, Western Blotting

Specifications
Troponin I, a.a. 87-93 | |
Unconjugated | |
Bovine cardiac Troponin I. | |
RUO | |
Primary | |
Semi-purified |
Monoclonal | |
Mouse | |
100 μg | |
Endocrine/Metabolism | |
Bovine, Chicken, Fish, Frog, Human, Mouse, Rabbit | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction