Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troponin I Mouse anti-Bovine, Chicken, Human, Mouse, Rabbit, Fish, Frog, Clone: C5, MilliporeSigma™

Catalog No. MAB1691MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1691MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1691MI Supplier MilliporeSigma Supplier No. MAB1691
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Troponin I using this Anti-Troponin I Antibody, clone C5 validated for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Troponin I
Applications ELISA, Western Blot
Classification Monoclonal
Clone C5
Conjugate Unconjugated
Formulation Purified immunoglobulin from Protein A Sepharose. Liquid in PBS pH 7.4 with 0.1% sodium azide as a preservative.
Gene Accession No. NM_000363.4
Gene Symbols TNNI3, TNNC1, CMH7, MGC116817, cTnI
Host Species Mouse
Immunogen Bovine cardiac Troponin I.
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Chicken, Fish, Frog, Human, Mouse, Rabbit
Content And Storage 2-8°C for up to 6 months.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.