Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRP1/TYRP1 Mouse anti-Mouse, Human, Clone: TA99, MilliporeSigma™

Catalog No. MABC592MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC592MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC592MI Supplier MilliporeSigma Supplier No. MABC592
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-TRP1/TYRP1 Antibody, clone TA99, Azide Free is validated for use in western blotting, immunohistochemistry, blocking of inteferring antibodies, immunoprecipitation and flow cytometry for the detection of TRP1/TYRP1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRP1/TYRP1
Applications Blocking Assay, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone TA99
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing PBS without preservatives.
Gene Accession No. NP_000541
Gene Symbols TYRP1, CAS2, TYRP, TYRRP
Host Species Mouse
Immunogen Whole cells corresponding to human TRP1/TYRP1.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.