Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

transient receptor potential cation channel, subfamily A, member 1, Mouse, Clone: 6G8, Abnova™

Catalog No. 89014231 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-231 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-231 Supplier Abnova Corporation Supplier No. H00008989M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant TRPA1.

The structure of the protein encoded by this gene is highly related to both the protein ankyrin and transmembrane proteins. The specific function of this protein has not yet been determined; however, studies indicate the function may involve a role in signal transduction and growth control. [provided by RefSeq

Sequence: IPNADKSLEMEILKQKYRLKDLTFLLEKQHELIKLIIQKMEIISETEDDDSHCSFQDRFKKEQMEQRNSRWNTVLRAVKAKTHHL

Specifications

Antigen transient receptor potential cation channel, subfamily A, member 1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6G8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant TRPA1.
Formulation PBS with no preservative; pH 7.4
Gene TRPA1
Gene Accession No. NM_007332
Gene Alias ANKTM1
Gene Symbols TRPA1
Host Species Mouse
Immunogen TRPA1 (NP_015628, 1033 a.a. ∼ 1117 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8989
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.