Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

transient receptor potential cation channel, subfamily C, member 3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89018849 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-849 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-849 Supplier Abnova Corporation Supplier No. H00007222A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant TRPC3.

Sequence: TARFLAFLQATKAQQYVDSYVQESDLSEVTLPPEIQYFTYARDKWLPSDPQ

Specifications

Antigen transient receptor potential cation channel, subfamily C, member 3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant TRPC3.
Formulation 50% glycerol
Gene TRPC3
Gene Accession No. NM_003305
Gene Alias TRP3
Gene Symbols TRPC3
Host Species Mouse
Immunogen TRPC3 (NP_003296, 485 a.a. ∼ 535 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7222
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.