Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TrpE Tag Monoclonal Antibody (17), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA112688
Description
TrpE is a commonly used tag in some protein expression systems. TrpE antibodies can be used for analysis of expression of proteins alone or in conjunction with protein specific antibodies. They can also be used as a negative control, as they do not react with any known mammalian protein.
Specifications
TrpE Tag | |
Monoclonal | |
Unconjugated | |
Mouse | |
Protein A/G | |
RUO | |
Tag | |
Antibody | |
IgG |
Western Blot | |
17 | |
PBS with 0.08% sodium azide | |
Full-length recombinant recombinant TrpE | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Tag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction