Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPM1 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB1556050UL
Description
Detect TRPM1 using this Anti-TRPM1 Antibody validated for use in western blotting.
Specifications
TRPM1 | |
Polyclonal | |
Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.05% sodium azide as a preservative. Reconstitute with 50μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min). | |
Rabbit | |
Affinity Purified | |
Primary | |
Human |
Western Blot | |
Unconjugated | |
TRPM1; Melastatin-1; MLSN1; LTRPC1 | |
Purified peptide from the C-terminus of human TRPM1 (Accession number O75560). | |
50 μL | |
NM_002420.4 | |
Lyophilized: −20°C for up to 12 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction