Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPM8 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
$204.00 - $482.50
Specifications
Antigen | TRPM8 |
---|---|
Concentration | 1.0 mg/ml |
Dilution | Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:300, KnockDown Validated |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP197311SS
|
Novus Biologicals
NBP197311SS |
0.025 mL |
Each for $204.00
|
|
|||||
NBP197311
|
Novus Biologicals
NBP197311 |
0.1 mL |
Each for $482.50
|
|
|||||
Description
TRPM8 Polyclonal specifically detects TRPM8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
TRPM8 | |
Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:300, KnockDown Validated | |
Polyclonal | |
Rabbit | |
Cancer, Tumor Suppressors | |
PBS (pH 7.4) with 0.05% Sodium Azide | |
Long transient receptor potential channel 6, LTrpC6, LTrpC-6, MGC2849, short form of the TRPM8 cationic channel, transient receptor potential cation channel, subfamily M, member 8, Transient receptor potential p8, transient receptor potential subfamily M member 8, trp-p8, TRPP8transient receptor potential cation channel subfamily M member 8 | |
TRPM8 | |
IgG | |
Affinity Purified |
1.0 mg/ml | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
Q7Z2W7, Q8R4D5, Q8R455 | |
79054 | |
A synthetic peptide made to an internal portion of the human TRPM8 protein (between residues 250-300) [UniProt# Q7Z2W7] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title