Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPML3 Mouse anti-Human, Mouse, Rat, Clone: N268/19, MilliporeSigma™

Catalog No. MABN667MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN667MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN667MI Supplier MilliporeSigma Supplier No. MABN667

Mouse Monoclonal Antibody

Detect Mucolipin-3 using this mouse monoclonal antibody, Anti-TRPML3 Antibody and clone N268/18 validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPML3
Applications Western Blot
Classification Monoclonal
Clone N268/19
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001240622
Gene Symbols MCOLN3
Host Species Mouse
Immunogen Linear peptide corresponding to the N-terminus of human TRPML3.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.