Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

transient receptor potential cation channel, subfamily V, member 1, Mouse, Clone: 1F5, Abnova™

Catalog No. 89019157 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-019-157 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-019-157 Supplier Abnova Corporation Supplier No. H00007442M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant TRPV1.

Capsaicin, the main pungent ingredient in hot chili peppers, elicits a sensation of burning pain by selectively activating sensory neurons that convey information about noxious stimuli to the central nervous system. The protein encoded by this gene is a receptor for capsaicin and is a nonselective cation channel that is structurally related to members of the TRP family of ion channels. This receptor is also activated by increases in temperature in the noxious range, suggesting that it functions as a transducer of painful thermal stimuli in vivo. Four transcript variants encoding the same protein, but with different 5' UTR sequence, have been described for this gene. [provided by RefSeq]

Sequence: CPDPLDGDPNSRPPPAKPQLSTAKSRTRLFGKGDSEEAFPVDCPHEEGELDSCPTITVSPVITIQRPGDGPTGARLLSQDSVAASTEKTLRLYDRRSIFEAVAQ

Specifications

Antigen transient receptor potential cation channel, subfamily V, member 1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1F5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant TRPV1.
Formulation PBS with no preservative; pH 7.4
Gene TRPV1
Gene Accession No. NM_080706
Gene Alias DKFZp434K0220/VR1
Gene Symbols TRPV1
Host Species Mouse
Immunogen TRPV1 (NP_542437, 21 a.a. ∼ 124 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7442
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.