Learn More
TRPV5, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human TRPV5 peptide using ARM Technology.
Supplier: Abnova Corporation H00056302K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene is a member of the transient receptor family and the TrpV subfamily. The calcium-selective channel encoded by this gene has 6 transmembrane-spanning domains, multiple potential phosphorylation sites, an N-linked glycosylation site, and 5 ANK repeats. This protein forms homotetramers or heterotetramers and is activated by a low internal calcium level. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
TRPV5 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
TRPV5 | |
TRPV5 | |
A synthetic peptide of human TRPV5 | |
RUO | |
56302 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
transient receptor potential cation channel, subfamily V, member 5 | |
TRPV5 | |
CAT2, ECAC1, OTRPC3 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.