Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Trypsin, Polyclonal

Catalog No. 6502971ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
65-029-71ML 1 mL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 65-029-71ML Supplier MilliporeSigma™ Supplier No. 6502971ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Trypsin Clone: in Human samples, and it is validated for ELISA, Immunoelectrophoresis, Immuno Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Trypsin
Applications ELISA, Immunoblot, Immunoblot, Immunoelectrophoresis
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS, pH 7.4.
Host Species Rabbit
Immunogen human pancreatic trypsin
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes cationic trypsin in pancreas tissue. Does not cross-react with other plasma proteins.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.