Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tryptase Mouse anti-Human, Clone: G3, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB1222
Description
This Anti-Tryptase Antibody, Mast Cell, clone G3 is validated for use in flow cytometry, immunohistochemistry, western blotting for the detection of Tryptase.
Specifications
Tryptase | |
Monoclonal | |
Unconjugated | |
NM_003294.3 | |
Mouse | |
100 μg | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, Western Blot | |
G3 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. (see product datasheet for specific buffer formulation) |
|
TPSAB1, tryptase-I, TPS2, Tryptase-1, TPSB1, TPS1, tryptase-III, EC 3.4.21.59, EC 3.4.21.59 | |
Protein G purfied | |
Primary | |
2-8°C | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction