Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tryptophan hydroxylase 2 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $426.00

Specifications

Antigen Tryptophan hydroxylase 2
Concentration 1.12 mg/mL
Dilution Western Blot 2 ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP177559SS
SDP
View Documents
Novus Biologicals
NBP177559SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NBP177559
SDP
View Documents
Novus Biologicals
NBP177559
0.1 mL
Each for $426.00
Only null left
Add to Cart
 
Description

Description

Tryptophan hydroxylase 2 Polyclonal specifically detects Tryptophan hydroxylase 2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Tryptophan hydroxylase 2
Western Blot 2 ug/ml
Polyclonal
Rabbit
Lipid and Metabolism
PBS and 30% Glycerol with 0.05% Sodium Azide
ADHD7, EC 1.14.16, EC 1.14.16.4, FLJ37295, MGC138872, Neuronal tryptophan hydroxylase, NTPHMGC138871, tryptophan 5-hydroxylase 2, Tryptophan 5-monooxygenase 2, tryptophan hydroxylase 2
TPH2
IgG
Affinity Purified
1.12 mg/mL
Western Blot
Unconjugated
RUO
Human
Q8IWU9
121278
A synthetic peptide made to an N-terminal portion of the human Tryptophan hydroxylase 2 protein (between residues 100-200) [UniProt# Q8IWU9]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.