Learn More
Tryptophan Hydroxylase Sheep anti-Human, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Tryptophan Hydroxylase |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified sheep polyclonal liquid in 150mM NaCl, 10mM HEPES (pH 7.5), 100μg/mL BSA and 50% glycerol. |
| Gene Symbols | TPH1; TPRH; MGC119994; TRPH; TPH; EC 1.14.16.4 |
| Host Species | Sheep |
| Immunogen | Recombinant rabbit tryptophan hydroxylase, isolated as inclusion bodies from E. coli and purified by preparative SDS-PAGE. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.