Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tryptophan Hydroxylase/Tyrosine Hydroxylase/Phenylalanine Hydroxylase Mouse anti-Human, Clone: PH8, MilliporeSigma™

Catalog No. MAB5278MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5278MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5278MI Supplier MilliporeSigma Supplier No. MAB5278
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Tryptophan Hydroxylase/Tyrosine Hydroxylase/Phenylalanine Hydroxylase Antibody, clone PH8 is validated for use in immunohistochemistry, immunoprecipitation, western blotting for the detection of Tryptophan Hydroxylase/Tyrosine Hydroxylase/Phenylalanine Hydroxylase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tryptophan Hydroxylase/Tyrosine Hydroxylase/Phenylalanine Hydroxylase
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone PH8
Conjugate Unconjugated
Formulation The immunoglobulin fraction has been purified from ascites fluid by Protein G chromatography. 500μg liquid protein at 2mg/mL in phosphate buffered saline (PBS). The product is supplied sterile-filtered through a 0.22 micron filter.
Gene Accession No. NM_004179.1
Gene Symbols TPH1, TPRH, MGC119994, TPH, TRPH, EC 1.14.16.4
Host Species Mouse
Quantity 500 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.