Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSC22D3, Mouse, Clone: 3A5, Abnova™

Catalog No. 89021359 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-359 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-359 Supplier Abnova Corporation Supplier No. H00001831M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant TSC22D3.

The protein encoded by this gene shares significant sequence identity with the murine TSC-22 and Drosophila shs, both of which are leucine zipper proteins, that function as transcriptional regulators. The expression of this gene is stimulated by glucocorticoids and interleukin 10, and it appears to play a key role in the anti-inflammatory and immunosuppressive effects of this steroid and chemokine. Transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq

Sequence: MAQSKLDCRSPVGLDCCNCCLDLAHRSGLQRGSSGENNNPGSPTVSNFRQLQEKLVFENLNTDKLNSIMRQDSLEPVLRDPCYLINEGICNRNIDQT

Specifications

Antigen TSC22D3
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3A5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant TSC22D3.
Formulation PBS with no preservative; pH 7.4
Gene TSC22D3
Gene Accession No. NM_198057
Gene Alias DIP/DKFZp313A1123/DSIPI/GILZ/TSC-22R/hDIP
Gene Symbols TSC22D3
Host Species Mouse
Immunogen TSC22D3 (NP_932174, 1 a.a. ∼ 97 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1831
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.