Learn More
TSC22D4, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human TSC22D4 peptide using ARM Technology.
Supplier: Abnova Corporation H00081628K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
TSC22D4 is a member of the TSC22 domain family of leucine zipper transcriptional regulators (see TSC22D3; MIM 300506) (Kester et al., 1999 [PubMed 10488076]; Fiorenza et al., 2001 [PubMed 11707329]).[supplied by OMIM
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
TSC22D4 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
TSC22D4 | |
TSC22D4 | |
A synthetic peptide of human TSC22D4 | |
RUO | |
81628 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
TSC22 domain family, member 4 | |
TSC22D4 | |
THG-1, THG1 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.