Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TSEN34 Ab-1, Polyclonal, Abnova

Catalog No. 89008152 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-008-152 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-008-152 Supplier Abnova Corporation Supplier No. H00079042B01

Mouse Polyclonal

tRNA splicing is a fundamental process required for cell growth and division. SEN34 is a subunit of the tRNA splicing endonuclease, which catalyzes the removal of introns, the first step in tRNA splicing (Paushkin et al., 2004).

Detection Antibody, Immunofluorescence, Western Blotting

Specifications

Antigen tRNA splicing endonuclease 34 homolog (S. cerevisiae)
Classification Polyclonal
Conjugate Unconjugated
Host Species Mouse
Immunogen TSEN34 (AAH00944.1, 1 a.a. ∽390 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Target Species Human
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.