Learn More
tRNA splicing endonuclease 34 homolog (S. cerevisiae), Mouse, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
tRNA splicing is a fundamental process required for cell growth and division. SEN34 is a subunit of the tRNA splicing endonuclease, which catalyzes the removal of introns, the first step in tRNA splicing (Paushkin et al., 2004 [PubMed 15109492]).[supplied by OMIM
Specifications
Specifications
| Antigen | tRNA splicing endonuclease 34 homolog (S. cerevisiae) |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a partial recombinant TSEN34. |
| Formulation | 50% glycerol |
| Gene | TSEN34 |
| Gene Accession No. | NM_024075 |
| Gene Alias | LENG5/PCH2C/SEN34/SEN34L |
| Gene Symbols | TSEN34 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.