Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSG101 [Tumor Susceptibility Gene 101] Mouse, Unlabeled, Clone: 51, BD

Catalog No. BDB612696 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612696 50 μg
BDB612697 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB612696 Supplier BD Biosciences Supplier No. 612696
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Tumor Susceptibility Gene 101 (TSG101) was identified in a random mutagenesis screen for potential tumor suppressors in NIH 3T3 cells. Altered transcripts of this gene have been detected in sporadic breast cancers and many other human malignancies. Though the function of TSG101 is not clearly understood, its protein structure includes motifs involved in transcription regulation, and TSG101 has been shown to modulate the activation of steroid hormone receptors. In addition, TSG101 may have a role in regulating ubiquination. The N-terminal region of TSG101 contains a domain that resembles the catalytically active region of ubiquitin conjugases. However, TSG101 lacks an active-site cysteine crucial to the function of these conjugases. Interestingly, TSG101 interferes with MDM2 ubiquination leading to a decrease in MDM2 decay and down-regulation of p53 protein. Thus, TSG101 may function as a dominant-negative inhibitor of ubiquination in pathways where protein expression is tightly regulated.

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen TSG101 [Tumor Susceptibility Gene 101]
Applications Western Blot
Classification Monoclonal
Clone 51
Concentration 250μg/mL
Conjugate Unconjugated
Description Tumor Susceptibility Gene 101
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human TSG101 aa. 229-319
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.