Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSLP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520321
Description
Despite its predicted molecular weight, TSLP often migrates at a higher molecular weight in SDS-PAGE. A suggested positive control is A-20 cell lysate. PA5-20321 can be used with blocking peptide PEP-0441.
TSLP is a 159 amino acid four helix-bundle hemopoietic cytokine that signals through a heterodimeric receptor complex consisting of the human TSLP receptor and the IL-7R alpha-chain. It impacts myeloid cells and induces the release of T cell-attracting chemokines from monocytes, enhances the maturation of CD11c (+) dendritic cells, naive CD4 (+), and CD8 (+) T cells into proallergic effectors, and primes dendritic cells to produce large amounts of IL-12 after CD40 ligand stimulation. TSLP influences the regulation of the positive selection of regulatory T cells and maintenance of peripheral CD4 (+) T cell homeostasis. It induces allergic inflammation by directly activating mast cells and may be involved in the pathophysiology of inflammatory arthritis. It is expressed in heart, liver and prostate.
Specifications
TSLP | |
Polyclonal | |
Unconjugated | |
TSLP | |
Thymic stroma-derived lymphopoietin; thymic stromal lymphopoietin; TSLP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
53603, 688621 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9JIE6 | |
TSLP | |
A 20 amino acid peptide from near the center of mouse TSLP. | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction