Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSLP Receptor Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520378
Description
Despite its predicted molecular weight, TSLP-R often migrates at a higher molecular weight in SDS-PAGE. A suggested positive control is human liver tissue lysate. PA5-20378 can be used with blocking peptide PEP-0495.
Thymic stromal lymphopoietin (TSLP) has recently been identified as an important factor capable of driving dendritic cell maturation and activation. It is involved in the positive selection of regulatory T cells, maintenance of peripheral CD4+ T cell homeostasis and the induction of CD4+ T cell-mediated allergic reaction. TSLP is also capable of supporting the growth of fetal liver and adult B cell progenitors and their differentiation to the IgM-positive stage of B cell development. Its receptor TSLP-R will bind TSLP in a low-affinity fashion in transfected cells; co-transfection with IL-7R-alpha cDNA results in high-affinity binding and a functional heteromeric complex. This heteromeric receptor requires stat5 for TSLP-mediated signal transduction and is inhibited by SOCS-1.
Specifications
TSLP Receptor | |
Polyclonal | |
Unconjugated | |
CRLF2 | |
CRL2; Crlf2; CRLF2Y; Crlm2; CRLM-2; cytokine receptor CRL2 precusor; Cytokine receptor-like 2; Cytokine receptor-like factor 2; cytokine receptor-like molecule 2; delta 1; ILXR; IL-XR; Ly114; lymphocyte antigen 114; P2RY8/CRLF2 fusion; thymic stromal lymphopoietin protein receptor; thymic stromal-derived lymphopoietin receptor; thymic stromal-derived lymphopoietin, receptor; Tpte2; transmembrane phosphoinositide 3-phosphatase and tensin homolog 2; TSLP receptor; Tslpr; type I cytokine receptor delta 1; WI2-87105E3.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64109 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9HC73 | |
CRLF2 | |
A 19 amino acid peptide from near the center of human TSLP receptor. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction