Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TSPAN2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7876
This item is not returnable.
View return policy
Description
TSPAN2 Monoclonal specifically detects TSPAN2 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
TSPAN2 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
O60636 | |
TSPAN2 | |
NS0 mouse myeloma cell line transfected with human TSPAN2 Met1-Ile221 Accession # O60636 | |
100 μg | |
Primary | |
Detects human TSPAN2 in ELISAs. | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
822509 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLJ12082, NET3, new EST tetraspan 3, tetraspan 2, Tetraspan NET-3, tetraspan TM4SF, tetraspanin 2, tetraspanin 2 isoform, tetraspanin-2, TSN2, Tspan-2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10100 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction