Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tau tubulin kinase 2, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016621 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-621 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-621 Supplier Abnova Corporation Supplier No. H00146057B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human TTBK2 protein.

This gene encodes a serine-threonine kinase that putatively phosphorylates tau and tubulin proteins. Mutations in this gene cause spinocerebellar ataxia type 11 (SCA11); a neurodegenerative disease characterized by progressive ataxia and atrophy of the cerebellum and brainstem. [provided by RefSeq

Sequence: MSGGGEQLDILSVGILVKERWKVLRKIGGGGFGEIYDALDMLTRENVALKVESAQQPKQVLKMEVAVLKKLQGKDHVCRFIGCGRNDRFNYVVMQLQGRNLADLRRSQSRGTFTISTTLRLGRQILESIESIHSVGFLHRDIKPSNFAMGRFPSTCRKCYMLDFGLARQFTNSCGDVRPPRAVAGFRGTVRYASINAHRNREMGRHDDLWSLFYMLVEFVVGQLPWRKIKDKEQVGSIKERYDHRLMLKHLPPEFSIFLDHISSLDYFTKPDYQLLTSVFDNSIKTFGVIESDPFDWEKTGNDGSLTTTTTSTTPQLHTRLTPAAIGIANATPIPGDLLRENTDEVFPDEQLSDGENGIPVGVSPDKLPGSLGHPRPQEKDVWEEMDANKNKIKLGICKAATEEENSHGQANGLLNAPSLGSPIRVRSEITQPDRDIPLVRKLRSIHSFELEKRLTLEPKPDTDKFLETWYKIVYFSF

Specifications

Antigen tau tubulin kinase 2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human TTBK2 protein.
Formulation PBS with no preservative; pH 7.4
Gene TTBK2
Gene Accession No. BC041876.1
Gene Alias KIAA0847/SCA11/TTBK
Gene Symbols TTBK2
Host Species Mouse
Immunogen TTBK2 (AAH41876.1, 1 a.a. ∼ 478 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Kinases
Primary or Secondary Primary
Gene ID (Entrez) 146057
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.