Learn More
tetratricopeptide repeat domain 8, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human TTC8 protein.
Supplier: Abnova Corporation H00123016D01P
Description
This gene encodes a protein that has been directly linked to Bardet-Biedl syndrome. The primary features of this syndrome include retinal dystrophy, obesity, polydactyly, renal abnormalities and learning disabilities. Experimentation in nonhuman eukaryotes suggests that this gene is expressed in ciliated cells and that it is involved in the formation of cilia. Alternate transcriptional splice variants have been characterized. [provided by RefSeq
Sequence: MSSEMEPLLLAWSYFRRRKFQLCADLCTQMLEKSPYDQAAWILKARALTEMVYIDEIDVDQEGIAEMMLDENAIAQVPRPGTSLKLPGTNQTGGPSQAVRPITQAGRPITGFLRPSTQSGRPGTMEQAIRTPRTAYTARPITSSSGRFVRLGTASMLTSPDGPFINLSRLNLTKYSQKPKLAKALFEYIFHHENDVKTALDLAALSTEHSQYKDWWWKVQIGKCYYRLGMYREAEKQFKSALKQQEMVDTFLYLAKVYVSLDQPVTALNLFKQGLDKFPGEVTLLCGIARIYEEMNNMSSAAEYYKEVLKQDNTHVEAIACIGSNHFYSDQPEIALRFYRRLLQMGIYNGQLFNNLGLCCFYAQQYDMTLTSFERALSLAENEEEAADVWYNLGHVAVGIGDTNLAHQCFRLALVNNNNHAEAYNNLAVLEMRKGHVEQARALLQTASSLAPHMYEPHFNFATISDKIGDLQRSYVAAQKSEAAFPDHVDTQHLIKQLRQHFAMLSpecifications
tetratricopeptide repeat domain 8 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human TTC8 protein. | |
TTC8 | |
BBS8 | |
Rabbit | |
Affinity Purified | |
RUO | |
123016 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_198309.2 | |
TTC8 | |
TTC8 (NP_938051.1, 1 a.a. ∼ 505 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.