Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTF-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 07601MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-601-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-601-MI Supplier MilliporeSigma Supplier No. 07601

Rabbit Polyclonal Antibody

Anti-TTF-1 Antibody is a high quality Rabbit Polyclonal Antibody for the detection of TTF-1 and has been validated in immunoprecipitation, western blotting and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTF-1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation rabbit antiserum containing 0.05% sodium azide before the addition of glycerol to 30%
Gene Symbols TTF1; TTF-I; NKX2A; TITF1
Host Species Rabbit
Immunogen 6His fusion protein corresponding to full-length TTF-1 (Thyroid transcription factor 1, also known as Thyroid nuclear factor 1, Homeobox protein Nkx-2.1)
Purification Method Serum
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_003317.3
Target Species Human, Mouse, Rat
Content And Storage −20°C for 24 months
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.