Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

transcription termination factor, RNA polymerase II, Mouse, Clone: 3D11, Abnova™

Catalog No. 89001081 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-081 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-081 Supplier Abnova Corporation Supplier No. H00008458M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant TTF2.

This gene encodes a member of the SWI2/SNF2 family of proteins, which play a critical role in altering protein-DNA interactions. The encoded protein has been shown to have dsDNA-dependent ATPase activity and RNA polymerase II termination activity. This protein interacts with cell division cycle 5-like, associates with human splicing complexes, and plays a role in pre-mRNA splicing. [provided by RefSeq

Sequence: EEVRCPEHGTFCFLKTGVRDGPNKGKSFYVCRADTCSFVRATDIPVSHCLLHEDFVVELQGLLLPQDKKEYRLFFRCIRSKAEGKRWCGSIPWQDPDSK

Specifications

Antigen transcription termination factor, RNA polymerase II
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3D11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant TTF2.
Formulation PBS with no preservative; pH 7.4
Gene TTF2
Gene Accession No. NM_003594
Gene Alias HuF2
Gene Symbols TTF2
Host Species Mouse
Immunogen TTF2 (NP_003585, 2 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 8458
Target Species Human, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.