Learn More
MilliporeSigma™ anti-Tubulin, δ 2 ; Polyclonal; ™ (Chemicon™)
Supplier: MilliporeSigma™ AB3203/DEL
Description
Tubulin, the dimeric subunit of microtubules, is submitted to post translational modifications including cyclic tyrosine removal and addition at the COOH terminus of the α subunit. The cycle involves two enzymes, TTL (Tubuline Tyrosine Ligase) and Tubulin carboxypeptidase, and generates two major forms: tyrosinated tubulin and detyrosinated tubulin (Glu-tubulin). Removal of the COOH-terminal Glu residue from the α chain of Glu-tubulin generates the δ2-tubulin form.
Electron Microscopy, Immunocytochemistry, Immunohistochemistry, Western Blotting
Specifications
| Tubulin, δ-2 | |
| Unconjugated | |
| Peptide corresponding to the C-terminal of δ 2 tubulin. | |
| 50 μg | |
| Cell Structure | |
| Echinodermata, Mammalia, Plant, Sea urchin |
| Polyclonal | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.