Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tubulin Rabbit anti-Mammals, Green Plants, Echinoderms, Sea Urchin, Polyclonal, MilliporeSigma™

Catalog No. AB3203MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3203MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3203MI Supplier MilliporeSigma Supplier No. AB3203
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Tubulin Antibody, δ 2 is validated for use in immunohistochemistry (paraffin), western blot, immunocytochemistry, immunohistochemistry for the detection of Tubulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tubulin
Applications Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in PBS.
Gene Symbols TUBD1; FLJ12709; delta-tubulin; Delta-tubulin; TUBD
Host Species Rabbit
Immunogen Peptide corresponding to the C-terminal of delta 2 tubulin.
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_016261.2
Target Species Echinodermata, Mammalia, Plant, Sea urchin
Content And Storage 2-8°C in undiluted aliquots for up to 6 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.