Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Β-Tubulin Mouse anti-Bovine, Human, Mouse, Rat, AlexaFluor™488, Clone: AA2, MilliporeSigma™

Catalog No. 16230MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-230-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-230-MI Supplier MilliporeSigma Supplier No. 16230
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-β-Tubulin Antibody, clone AA2, Alexa Fluor 488 conjugate is validated for use in flow cytometry, immunofluoresence, western blot, IC for the detection of β-Tubulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen β-Tubulin
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone AA2
Conjugate Alexa Fluor 488
Gene Accession No. NM_
Gene Symbols LOC647000
Host Species Mouse
Immunogen Purified bovine brain tubulin; epitope has been mapped to amino acids 412-430
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Rat
Content And Storage 4°C for one year
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.