Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Tubulin Mouse anti-Human, Rat, Guinea Pig, Bovine, Mouse, Pig, Avian, AlexaFluor™488, Clone: DM1A, MilliporeSigma™

Catalog No. 16232MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-232-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-232-MI Supplier MilliporeSigma Supplier No. 16232
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-α-Tubulin Antibody, clone DM1A, Alexa Fluor 488 conjugate is an antibody against α-Tubulin for use in flow cytometry, immunocytochemistry, immunofluorescence and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen α-Tubulin
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone DM1A
Conjugate Alexa Fluor 488
Formulation Alexa Fluor™ 488 conjugated, purified mouse monoclonal IgG1 in buffer containing PBS containing 1% BSA, 0.05% Tween™-20, 0.05% sodium azide.
Liquid at 2-8°C.
Gene Accession No. NP_006000
Gene Symbols TUBA1A, B-ALPHA-1, FLJ25113, hum-a-tub1, TUBA3, hum-a-tub2
Host Species Mouse
Immunogen Purified human brain α-Tubulin. The epitope recognized by this antibody has been mapped to amino acids 426-450 (AALEKDYEEVGVDSV-EGEGEEEGEE).
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Avian, Bovine, Guinea Pig, Human, Mouse, Pig, Rat
Content And Storage 2-8°C for 12 months. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.