Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tubulin Β Mouse anti-Bovine, Human, Rat, Clone: 5H1, MilliporeSigma™

Catalog No. MAB380MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB380MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB380MI Supplier MilliporeSigma Supplier No. MAB380
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Tubulin Antibody, β, clone 5H1 is an antibody against Tubulin for use in immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tubulin β
Applications Immunohistochemistry
Classification Monoclonal
Clone 5H1
Conjugate Unconjugated
Formulation Ascites fluid containing sodium azide. Liquid
Gene Accession No. NM_006086.2
Gene Symbols TUBB3, TUBB4, MC1R, beta-4
Host Species Mouse
Immunogen Rat tubulin.
Quantity 100 μL
Primary or Secondary Primary
Target Species Bovine, Human, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.