Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tubulin Mouse anti-All, Human, Mouse, Rat, Clone: KMX-1, MilliporeSigma™

Catalog No. MAB3408MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3408MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3408MI Supplier MilliporeSigma Supplier No. MAB3408
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Tubulin Antibody, beta, clone KMX-1 is validated for use in immunocytochemistry, western blotting for the detection of Tubulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tubulin
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone KMX-1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2b liquid in buffer containing 0.02M phosphate buffer, 0.25M NaCl with 0.1% sodium azide.
Gene Accession No. NM_006086.2
Gene Symbols TUBB3, TUBB4, MC1R, beta-4
Host Species Mouse
Immunogen Tubulin purified from polycephalum myxamoebae
Purification Method Protein A purfied
Quantity 50 μg
Primary or Secondary Primary
Target Species All species, Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.