Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tubulin Mouse anti-Avian, Bovine, Human, Mouse, Monkey, Pig, Rat, Sheep, Clone: TU-20 (Similar to TUJ1), MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB1637
Description
Anti-Tubulin Antibody, beta III isoform, C-terminus, clone TU-20 (Similar to TUJ1) is an antibody against Tubulin for use in ELISA, immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin), immunoprecipitation and western blotting with more than 55 product citations.
Specifications
| Tubulin | |
| Monoclonal | |
| Unconjugated | |
| NM_006086.2 | |
| Mouse | |
| Unpurified | |
| Primary | |
| −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
| IgG1 |
| ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| TU-20 (Similar to TUJ1) | |
| Mouse monoclonal IgG1 ascites in buffer containing PBS containing 1% BSA and 15mM sodium azide. | |
| TUBB3, TUBB4, beta-4, MC1R | |
| A synthetic peptide corresponding to amino acids 443-450 (ESESQGPK) of human class III beta-tubulin conjugated to KLH. | |
| 100 μL | |
| Avian, Bovine, Human, Monkey, Mouse, Pig, Rat, Sheep | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction