Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tubulin Rat anti-Human, Mouse, Rat, Clone: YL1/2, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma MAB1864
Description
Detect Tubulin using this Anti-Tubulin Antibody, clone YL1/2 validated for use in ELISA, immunohistochemistry, immunoprecipitation, RIA and western blotting.
Specifications
Tubulin | |
Monoclonal | |
Unconjugated | |
Rat | |
Protein A purfied | |
Primary | |
−20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. | |
IgG2a |
ELISA, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot | |
YL1/2 | |
Purified rat monoclonal IgG2a in buffer containing phosphate buffered saline (pH 7.4) with 0.09% sodium azide. | |
Yeast tubulin | |
100 μg | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction