Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tubulin Β II Mouse anti-Mammals, MilliporeSigma™

Catalog No. MAB5562MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5562MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5562MI Supplier MilliporeSigma Supplier No. MAB5562

Mouse Monoclonal Antibody

Detect Tubulin using this Anti-Tubulin Antibody, β II validated for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tubulin β II
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Liquid. Contains no preservative.
Gene Accession No. NM_006086.2
Gene Symbols TUBB3, TUBB4, MC1R, beta-4
Host Species Mouse
Immunogen Synthetic peptide EEEGEDEA.
Quantity 100 μL
Primary or Secondary Primary
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.