Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tubulin Β-III Mouse anti-Bovine, Human, Primate, Mouse, Pig, Rat, AlexaFluor™488, Clone: TU-20, MilliporeSigma™

Catalog No. CBL412XMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
40 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL412XMI 40 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL412XMI Supplier MilliporeSigma Supplier No. CBL412X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Tubulin β-III using this Anti-Tubulin β-III Antibody, clone TU-20, Alexa Fluor™488 Conjugated validated for use in immunocytochemistry, immunofluoresence, western blot, immunohistochemistry and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tubulin β-III
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone TU-20
Conjugate Alexa Fluor 488
Formulation Purified immunoglobulin in 0.02M Phosphate buffer, 0.25M NaCl containing 0.05% sodium azide.
Gene Accession No. NP_001184110
Gene Symbols MC1R, TUBB4, beta-4
Host Species Mouse
Purification Method Protein G purfied
Quantity 40 μL
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Pig, Primate, Rat
Content And Storage 2-8°C for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.