Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tubulin Β III Mouse anti-Rat, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB5564
Description
Anti-Tubulin Antibody, β III detects level of Tubulin and has been published and validated for use in western blot, immunohistochemistry.
Specifications
Tubulin β III | |
Monoclonal | |
Liquid in 0.02M phosphate buffer, 0.25MNaCl, pH 7.6 with 0.1% sodium azide.. | |
TUBB3, TUBB4, MC1R, beta-4 | |
100 μg | |
Rat | |
Purified |
Immunohistochemistry, Western Blot | |
Unconjugated | |
NM_006086.2 | |
Mouse | |
Primary | |
2-8°C in undiluted aliquots for up to 6 months. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction