Learn More
Tumor Necrosis Factor-α Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Tumor Necrosis Factor-α |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity Purified Immunoglobulin. Lyophilized from PBS pH 7.2. Contains no preservative. Reconstitute with sterile distilled water. |
| Gene Symbols | TNF; TNFSF2; Cachectin; cachectin; DIF; TNF-alpha; OTTHUMP00000037669; TNFA; TNF-a |
| Host Species | Rabbit |
| Immunogen | Highly purified (≥98%) recombinant rat TNF-alpha |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.