Learn More
Tumor Necrosis Factor-α Rabbit anti-Mouse, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Tumor Necrosis Factor-α |
| Applications | ELISA, Neutralization, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Immunoglobulin purified by affinity chromatography employing immobilized mTNF-alpha matrix. A sterile filtered antibody solution (1mg/mL) was lyophilized from 0.5X PBS, pH 7.2. Reconstitute with sterile distilled water. |
| Gene Symbols | TNF; TNFSF2; Cachectin; cachectin; DIF; TNF-alpha; OTTHUMP00000037669; TNFA; TNF-a |
| Host Species | Rabbit |
| Immunogen | Highly pure (≥98%) recombinant Mouse TNF-alpha. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.