Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tumor Necrosis Factor-Β Mouse anti-Human, Clone: 9B9, MilliporeSigma™

Catalog No. MAB1098MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1098MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1098MI Supplier MilliporeSigma Supplier No. MAB1098
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Tumor Necrosis Factor-β Antibody, clone 9B9 detects level of Tumor Necrosis Factor-β and has been published and validated for use in immunocytochemistry, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tumor Necrosis Factor-β
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 9B9
Conjugate Unconjugated
Formulation Purified immunoglobulin (Protein A-agarose chormotography). One bottle of lyophilized preparation contains:200ug mouse IgG, 2 mg potassium phosphate buffer, 2.9 mg sodium chloride, and 20 mg raffinose.Reconstitution with 1 ml sterile, distilled water res
Gene Accession No. NM_000595.2
Gene Symbols LTA, OTTHUMP00000037612, TNF-beta, OTTHUMP00000037613, LT-alpha, LT, TNFB, TNFSF1
Host Species Mouse
Immunogen Human, recombinant tumor necrosis factor-beta (TNF-beta, lymphotoxin) produced in E. Coli.
Quantity 200 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.