Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TWEAK Mouse anti-Human, NA/LE, Unlabeled, Clone: CARL-1, BD

Catalog No. BDB562203 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562203 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562203 Supplier BD Biosciences Supplier No. 562203

Mouse Monoclonal Antibody

The CARL-1 monoclonal antibody specifically binds to CD255, also known as TWEAK (TNF-related WEAK inducer of apoptosis). The CD255 cytokine belongs to the tumor necrosis factor (TNF) ligand family and is encoded by the TNFSF12 ( Tumor necrosis factor ligand superfamily member 12 ) gene. CD255 (TWEAK) is present as a type II transmembrane protein form and a smaller, biologically-active secreted isoform. CD255 is expressed on most tissues and non-lymphoid tumor cell lines but has weak or no expression on hematopoietic lineage cell lines. CD255 is not detectable on resting peripheral blood mononuclear cells. However, its expression is upregulated on peripheral blood monocytes in response to IFN- γstimulation. CD255 can act through the TWEAK Receptor (aka, TWEAK-R, TNFRSF12A, Fn14, and CD266) to induce a variety of target cellular responses including NF-kB activation, proliferation, apoptosis, IL-8 and GM-CSF production. Clone CARL-2, a different monoclonal antibody, also binds to TWEAK although it binds to a different TWEAK epitope.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TWEAK
Applications Flow Cytometry
Classification Monoclonal
Clone CARL-1
Concentration 1.0mg/mL
Conjugate Unconjugated
Description CD255; TWEAK; TNFSF12; APO3 ligand; APO3L; DR3LG
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Host Species Mouse
Immunogen Human Transfected Cell Line
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.