Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

twinfilin, actin-binding protein, homolog 1 (Drosophila), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018408 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-408 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-408 Supplier Abnova Corporation Supplier No. H00005756D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human TWF1 protein.

This gene encodes twinfilin, an actin monomer-binding protein conserved from yeast to mammals. Studies of the mouse counterpart suggest that this protein may be an actin monomer-binding protein, and its localization to cortical G-actin-rich structures may be regulated by the small GTPase RAC1. [provided by RefSeq

Sequence: MTRRGRRRSSHFLGPPAGAAGCTQRRSRELAAAAMSHQTGIQASEDVKEIFARARNGKYRLLKISIENEQLVIGSYSQPSDSWDKDYDSFVLPLLEDKQPCYILFRLDSQNAQGYEWIFIAWSPDHSHVRQKMLYAATRATLKKEFGGGHIKDEVFGTVKEDVSLHGYKKYLLSQSSPAPLTAAEEELRQIKINEVQTDVGVDTKHQTLQGVAFPISREAFQALEKLNNRQLNYVQLEIDIKNEIIILANTTNTELKDLPKRIPKDSARYHFFLYKHSHEGDYLESIVFIYSMPGYTCSIRERMLYSSCKSRLLEIVERQLQMDVIRKIEIDNGDELTADFLYEEVHPKQHAHKQSFAKPKGPAGKRGIRRLIRGPAETEATTD

Specifications

Antigen twinfilin, actin-binding protein, homolog 1 (Drosophila)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human TWF1 protein.
Formulation PBS with no preservative; pH 7.4
Gene TWF1
Gene Accession No. NM_002822.3
Gene Alias A6/MGC23788/MGC41876/PTK9
Gene Symbols TWF1
Host Species Rabbit
Immunogen TWF1 (NP_002813.2, 1 a.a. ∼ 384 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Kinases
Primary or Secondary Primary
Gene ID (Entrez) 5756
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.