Learn More
Twist1 (Twist-related protein 1) Rabbit anti-Human, Mouse, Canine, Pig, Rhesus Macaque, Chimpanzee, Horse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABD29
Description
Detect Twist1 (Twist-related protein 1) using this Anti-Twist1 (Twist-related protein 1) Antibody validated for use in immunohistochemistry (paraffin) and immunocytochemistry.
Specifications
Twist1 (Twist-related protein 1) | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Canine, Chimpanzee, Equine, Human, Mouse, Pig, Rhesus Monkey |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
TWIST1; BHLHA38; TWIST | |
KLH-conjugated linear peptide corresponding to human Twist1 near the N-terminus. | |
100 μg | |
NP_000465 | |
2-8°C for 12 months |